Environmental Express Oakton pH Calibration Buffer Pouches, Assorted; 20/Pk

Representative image only.

Environmental Express Oakton pH Calibration Buffer Pouches, Assorted; 20/Pk

On-the-go convenient pH calibration

₹7,900.00INR / Pkg of 20

₹7,900.00INR / Pkg of 20

Buy It Together

Current Item

Environmental Express Oakton pH Calibration Buffer Pouches, Assorted; 20/Pk
Environmental Express Oakton pH Calibration Buffer Pouches, Assorted; 20/Pk
3565304

₹7,900.00INR / Pkg of 20
+ + + + +
Jenway 025 157 Redox Standard, 200 mV; 500 mL
7900029

₹16,200.00INR / Each
+ + + +
Environmental Express Electrode Storage Solution; 500 mL
0065304

₹3,700.00INR / Pint of 1
=
Total Price 185300.00 ADD 10 ITEMS TO CART
Product Description

Includes

Five pouches each of pH 4, 7, and 10 buffers and rinse solution.

Key Features

  • Single-use pouch—just tear, dip, calibrate
  • Available in common pH values 4, 7, and 10
  • Standardized to NIST-traceable pH references
  • Pack contains 20 pouches
  • Contain buffer pouches 4, 7, and 10 – Most commonly used for three-point calibration
  • 1 year warranty

More About this Item

Buffers are traceable and offer ±0.01 accuracy at 77 °F (25 °C). Pack of 20 pouches.

Five pouches each of pH 4, 7, and 10 buffers and rinse solution. All Oakton pH buffer solutions are manufactured under ISO 9000 quality guidelines and traceable to NIST standard (for meters with 0.01 resolution). A pH/temperature table is printed on the label for accurate calibrations.

1 Year Warranty
Specifications
  • Buffer Kit Values4.01, 7.00, and 10.00
  • Volume (mL)20
  • DescriptionpH Calibration Buffer Pouches, Assorted; 20/Pk
  • Warranty1 year
Documents & Certificates
Reviews
Q&A