Environmental Express Oakton pH 700 Benchtop Meter with All-in-One pH Electrode

Representative image only.

Environmental Express Oakton pH 700 Benchtop Meter with All-in-One pH Electrode

Designed for today's crowded laboratory benchtops!

₹92,900.00INR / Each

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

₹92,900.00INR / Each

Buy It Together

Current Item

Environmental Express Oakton pH 700 Benchtop Meter with All-in-One pH Electrode
Environmental Express Oakton pH 700 Benchtop Meter with All-in-One pH Electrode
3541903

₹92,900.00INR / Each
+ + + + + + + + +
Environmental Express Electrode Storage Solution; 500 mL
0065304

₹3,700.00INR / Pint of 1
=
Total Price 644000.00 ADD 10 ITEMS TO CART
Product Description

Includes

meter, all-in-one pH electrode, electrode stand, universal 110/240 VAC power supply and adapters.

Key Features

  • Compact footprint is over 40% smaller than other benchtop meters
  • Large display permits easier readout viewing
  • 3 year warranty

More About this Item

The easy-to-use, microprocessor-based pH 700 benchtop meter measures pH, mV, relative mV, and temperature. The large, dual-display LCD provides excellent visibility—even from a distance. Automatic temperature compensation (ATC) maintains reading accuracy even with fluctuating temperatures. Five-point pH calibration, with automatic buffer recognition for both USA (pH 1.68, 4.01, 7.00, 10.01, 12.45) and NIST (pH 1.68, 4.01, 6.86, 9.18, 12.45) pH buffers, makes for easy calibration no matter which requirement you need to meet.

Meter features HOLD, ATC, °C measurement, recall of electrode slopand offset, and electrode diagnostics. The convenient pull-out quick reference card keeps procedures handy at all times. Meter accepts any pH or ORP electrodes with a BNC connector.

3 Year WarrantyConformite EuropeenneCanadian Standards Association
Specifications
  • Meter TypeBenchtop
  • Display typeLCD
  • ConnectionspH: BNC; Temperature: 2.5 mm
  • Min pH (pH)-2
  • Max pH (pH)16
  • pH Resolution0.01/0.1
  • pH Accuracy±0.01 pH
  • Buffer RecognitionNIST and USA
  • CalibrationUp to 5 points
  • Min mV (mV)-2000
  • Max mV (mV)2000
  • mV Resolution0.1 or 1 mV
  • mV Accuracy±0.2 mV or 0.05%, whichever is greater
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • Temperature compensationAutomatic or manual
  • Power (VAC)100 to 240
  • Power (Hz)50 or 60
  • Width (cm)17.4625
  • Height (cm)6.985
  • Width (in)6 7/8
  • Length (in)8 17/64
  • Height (in)2 3/4
  • DescriptionBenchtop Meter with "All-in-One" pH Electrode
  • Warranty3 year
Reviews
Q&A