Environmental Express EC 100 Portable Conductivity Meter Kit with Case, EC/ATC Probe, and Solutions

Representative image only.

Environmental Express EC 100 Portable Conductivity Meter Kit with Case, EC/ATC Probe, and Solutions

Environmental Express | Item: 3560415

Convenient, compact design with large display

₹82,500.00INR / Each

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

₹82,500.00INR / Each

Buy It Together

Current Item

Product Description

Includes

EC 100 portable conductivity meter, plastic conductivity electrode, temperature probe, 50 mL conductivity calibration solutions (84 μS, 141 μS, & 12.85 mS), and carrying case.

Key Features

  • Measured parameter and temperature shown on display at same time
  • Stability, calibration, and low-battery indicators
  • Waterproof and dust-resistant IP57 rating – great for use under harsh environments
  • Replacement Con/ATC probes 35606-55 K = 1.0, 35606-53 K = 0.1 and 35606-57 K = 10.0
  • 2 year warranty

More About this Item

The new addition to the Environmental Express Oakton product line features updates to measuring water quality easier and faster. The larger display is 2.75 in. by 2.0 in. for better viewing angle, and has indicators for battery life, stability, and calibration. It also shows both the parameter and temperature measurements at the same time. The EC 100 meter measures conductivity, TDS, salinity, and temperature.

The built-in microprocessor chip enables advanced functions, such as auto calibration, auto temperature compensation, parameter setting, self-diagnosis, auto power-off, and low-battery reminders. The meter uses advanced digital processing technology, intelligently improving the response time and accuracy of your measurements. Stable reading and auto lock display modes are available.

Two year warrantyConformite Europeenne
Specifications
  • Meter TypeConductivity, TDS, salinity, temperature
  • Display typeLCD
  • ConnectionsBNC & Φ2.5 socket
  • Buffer RecognitionAutomatic standard and user-defined conductivity calibration solutions
  • Calibration1-4 points automatic
  • Min Conductivity0uS/cm
  • Max Conductivity0.00 to 19.99 μS/cm, 20.0 to 199.9 μS/cm,200 to 1999 μS/cm; 2.00 to 19.99 mS/cm, 20.0 to 199.9 mS/cm
  • Conductivity Resolution0.01, 0.1, 1 μS/cm; 0.01, 0.1 mS/cm
  • Conductivity Accuracy±1.0% full scale
  • CellK=1.0
  • Cell Constant0.1, 1, 10 cm–1
  • Min TDS0 g/L
  • Max TDS100 μg/L
  • TDS Accuracy±1.0% full scale
  • Min Salinity0 ppt
  • Max Salinity100 ppt
  • Salinity Accuracy±1.0% full scale
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • Min Temperature (° F)32
  • Max Temperature (° F)212
  • Temperature Resolution0.1°F/°C
  • Temperature Accuracy±0.9°F (0.5°C) ±1 digit
  • Temperature compensation0 to 100˚C (manual or automatic)
  • Temperature Coefficient2.0%/°C
  • WaterproofIP57
  • Battery3 AA (included)
  • Battery life (hr)400
  • Length (cm)9.1
  • Width (cm)19
  • Height (cm)3.3
  • Width (in)7 31/64
  • Length (in)3 37/64
  • Height (in)1 19/64
  • Stirring ControlNo
  • NIST Calibration IncludedNo
  • Probe IncludedEC/temperature (ATC)
  • Accessories IncludedCarrying case and solutions
  • DescriptionOakton EC 100 Portable Conductivity Meter Kit with Case, EC and ATC probes, and Solutions
Reviews
Q&A