Environmental Express Oakton® Buffer Pack; 500 mL of each pH 4.01, 7.00, and 10.01 (3 x 500 mL each)

Representative image only.

Environmental Express Oakton® Buffer Pack; 500 mL of each pH 4.01, 7.00, and 10.01 (3 x 500 mL each)

Environmental Express | Item: 0594210

Save over 25% on pack compared to ordering separately

₹5,900.00INR / Pkg of 3

₹5,900.00INR / Pkg of 3

Buy It Together

Current Item

Environmental Express Oakton® Buffer Pack; 500 mL of each pH 4.01, 7.00, and 10.01 (3 x 500 mL each)
Environmental Express Oakton® Buffer Pack; 500 mL of each pH 4.01, 7.00, and 10.01 (3 x 500 mL each)
0594210

₹5,900.00INR / Pkg of 3
+
Environmental Express Zero Oxygen Calibration Solution, 500 mL
0065300

₹4,800.00INR / Pint of 1
+
Environmental Express Electrode Storage Solution; 500 mL
0065304

₹3,700.00INR / Pint of 1
+ + + + + + + =
Total Price 224100.00 ADD 10 ITEMS TO CART
Product Description

Includes

500-mL plastic bottle of each pH solution: 4.01, 7.00, and 10.01.

Key Features

  • Lab-grade buffers available in multiple volumes to reduce waste and support various levels of throughput
  • Color coded for easy identification in busy labs
  • Pack counts to meet a variety of testing schedules
  • Cleaning and storage solutions available to ensure better accuracy and stability
  • Most common "USA" pH calibration buffers in one convenient pack
  • 2 year warranty

More About this Item

Environmental Express pH Buffer Bottles have pH/temperature tables printed on the labels for accurate calibrations. Manufactured under ISO 9000 quality guidelines and traceable to NIST standards. If unopened, shelf life is 2 years; once opened, use pH 4 and pH 7 within 90 days, pH 10 within 30 days.

Store buffers away from direct sunlight.

Color coded for quick visual reference:
pH 4.01 buffer in red
pH 7.00 buffer in yellow
pH 10.01 buffer in blue

To obtain a Certificate of Analysis on each item within the pack, please visit each individual items' page and enter the lot number in the space provided. For example, type part number 00654-00 into the search box at the top web page, then scroll down on the page to the CERTIFICATE OF ANALYSIS box and enter the lot number in the space provided.

To obtain a Certificate of Analysis on each item within the pack, please visit each individual items' page and enter the lot number in the space provided. For example, type part number 00654-00 into the search box at the top web page, then scroll down on the page to the CERTIFICATE OF ANALYSIS box, and enter the lot number in the space provided.

Specifications
  • Buffer Kit Values4.01, 7.00, and 10.01
  • Volume (mL)500
  • DescriptionBuffer Pack; 500 mL of each pH 4.01, 7.00, and 10.01
Reviews
Q&A