Fytertech Spilfyter Wastewater Classifier Kit, 20 Strips, Gloves, Thread, Chart, Plastic Case
Representative image only.
Fytertech Spilfyter Wastewater Classifier Kit, 20 Strips, Gloves, Thread, Chart, Plastic Case
Fytertech | Mfr: 581020| Item: 8642418
Classify unknown spills in 30 seconds
₹59,400.00INR / Each
₹59,400.00INR / Each
Buy It Together
Current Item
Fytertech Spilfyter Wastewater Classifier Kit, 20 Strips, Gloves, Thread, Chart, Plastic Case
Fytertech Spilfyter Wastewater Classifier Kit, 20 Strips, Gloves, Thread, Chart, Plastic Case
8642418
₹59,400.00INR / Each
+
Fytertech Spilfyter Chemical test strips, case of 50
8642350
₹126,000.00INR / Case of 50
+
Fytertech Spilfyter Wastewater Reference Chart
8642356
₹4,200.00INR / Each
=
Total Price 189600.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Kit includes twenty test strips, two bobbins of thread, one color reference chart, one pair of nitrile gloves, and one plastic case. Additional test strips and charts may also be ordered separately.
More About this Item
Convenient strips change color to identify risk type and classify spill for proper treatment. Compare color indicator to laminated, reusable reference chart to classify unknown liquid.
Specifications
  • DescriptionWastewater Classifier Kit, 20 Strips, Gloves, Thread, Chart, Plastic Case