Fytertech Spilfyter Mercury Vapor Suppressor Powder, 1 lb Shaker Bottle, 5/Case
Representative image only.
Fytertech Spilfyter Mercury Vapor Suppressor Powder, 1 lb Shaker Bottle, 5/Case
Fytertech | Mfr: 460005| Item: 8642459
Absorb dangerous mercury vapors after a spill
₹41,000.00INR / Case of 5
₹41,000.00INR / Case of 5
Product Description
More About this Item
Vapor suppressor adsorbs the dangerous mercury vapors that can be difficult to remove using other absorbents. Can achieve an absorption capacity of 20 g of mercury/100 g of suppressor.