NPS Spilfyter Kolor-Safe Base Neutralizer, 1 gallon
Representative image only.
NPS Mfr # 430004 – Item # WW-86423-13
Three-color change lets you know when spill is completely neutralized
PRODUCT OPTIONS
₹9,900.00INR / Gallon of 1
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionBase Neutralizer, 1 gallon
More About this Item
Protect your workers and facility from base spills by quickly and efficiently neutralizing them. Simply apply the neutralizer until the spill goes through the three-color change. Base neutralizer changes from red to blue to yellow to indicate total neutralization.