Fytertech Spilfyter Grab & Go Hazmat Spill Kit, Absorbs 5 gallons, 1/Bag
Representative image only.
Fytertech Spilfyter Grab & Go Hazmat Spill Kit, Absorbs 5 gallons, 1/Bag
Fytertech | Mfr: 2053BG| Item: 8642408
Kit includes everything you need for quick spill clean up, in a handy zipper bag
₹21,700.00INR / Bag of 1
₹21,700.00INR / Bag of 1
Product Description
Includes
Kit includes three 3" x 4' socks, twenty 16" x 18" pads, one pair of nitrile gloves, one pair of goggles, one disposal bag with twist tie, and one clear zipper bag.
More About this Item
Hazmat sorbents are specifically designed to absorb aggressive fluids such as acids and bases. Ideal for laboratory use.
Specifications
  • Absorption5 gal. (19 L)
  • DescriptionHazmat Spill Kit, Absorbs 5 gallons, 1/Bag