Fytertech Spilfyter Grab & Go Acid/Base Bucket Spill Kit
Representative image only.
Fytertech Spilfyter Grab & Go Acid/Base Bucket Spill Kit
Fytertech | Mfr: 270000| Item: 8642449
Kit contents are packed in a 5-gallon bucket for easy portability and disposal of materials
₹44,200.00INR / Each
₹44,200.00INR / Each
Product Description
Includes
Three 3" x 4' hazmat socks, fifteen 12" x 12" hazmat pads, one quart liquid acid neutralizer, one quart liquid base neutralizer, one chemical classifier kit, pair of nitrile gloves, pair of goggles, disposal bag with twist tie, and 5-gallon plastic bucket.
Key Features
  • Pour liquid neutralizer on floors or spray onto vertical surfaces
  • Use chemical classifier to detect the presence of other chemicals
More About this Item
Grab & Go buckets contain everything you need to quickly clean-up and dispose of a spill. Kit includes materials to neutralize acids and bases.
Specifications
  • Absorption7 gal. (27 L)
  • DescriptionAcid/Base Bucket Spill Kit