Labconco 3923401 Formaldehyde Carbon Filter for FilterMate™ Portable Exhausters
Representative image only.
Labconco 3923401 Formaldehyde Carbon Filter for FilterMate™ Portable Exhausters
Labconco | Mfr: 3923401| Item: 0910152
Filter removes contaminants before returning air to room or exhausting to outside
₹147,000.00INR / Each
₹147,000.00INR / Each
Product Description
Key Features
  • Compatible with Labconco® FilterMate™ portable exhausters
  • Type CI
  • Contains 14 lb of impregrated carbon media
More About this Item
Optimize performance of your Labconco FilterMate™ portable exhausters with appropriate filters.
Treated carbon media is suitable for vapors from formaldehyde.
Specifications
  • Filter TypeFormaldehyde carbon
  • DescriptionFormaldehyde Carbon Filter for FilterMate™ Portable Exhausters