This item is now discontinued. We recommend the following as suggested replacements: WW-18100-23
Elma Tec Clean A4 Ultrasonic Cleaner Detergent, Universal; 10 L
Representative image only.
Elma Mfr # 800 0131 – Item # WW-59988-62
Biodegradable, concentrated cleaners with long shelf lives
  • Several formulas to suit various industrial cleaning applications
SPECS & DESCRIPTION
Specifications & Description
  • FormatLiquid
  • Volume (gallons)2.6
  • Volume (Liters)10
  • ApplicationsManual or ultrasonic cleaning. Industrial use.
  • DescriptionUltrasonic Cleaner Detergent, Universal; 10 L
Key Features
  • Several formulas to suit various industrial cleaning applications
More About this Item
A4 solution is a universal cleaner for removing oil, grease, and polishing media—suitable for nonferrous metals, glass, rubber, ceramics, plastics, and other alkaline-resistant materials.