AccuMac Sec. SPRT, 0.25" x 12" , -200~670C, Spade SKU 9045257

Representative image only.

AccuMac Sec. SPRT, 0.25" x 12" , -200~670C, Spade SKU 9045257

AccuMac | Item: 9045257

State-of-the-art, high-accuracy temperature probes at a fraction of the cost

₹705,000.00INR / Each

₹705,000.00INR / Each
Product Description

Key Features

  • Equivalent to Fluke Calibration / Hart Scientific Model 5628
  • 1 year warranty

More About this Item

AM1762 Series—Secondary standard SPRT/RTD probes: These probes fill the gap between affordable, but temperature limited secondary PRTs and more expensive, yet highly accurate SPRTs. The unique design makes them very stable both short term (±2 mK at TPW) and long term (<19 mK typical at TPW after 500 hours) accuracy is ±6 mK at TPW . These high performance probes fully meet ITS-90 criteria for reference thermometers and are an excellent choices for reference standards for metrology labs of various levels. The Am1762 is nominally 25.5 ohm

Probes may be available in different lengths and terminations than what is shown in the ordering table above: please contact our Application Specialists for special order options.

1 Year Warranty
Specifications
  • Probe TypeSecondary Reference PRT/RTD
  • Min Temperature (° F)-328
  • Max Temperature (° F)1238
  • Min Temperature (° C)-200
  • Max Temperature (° C)670
  • Temperature Accuracy±0.006°C and ±0.004°C/100 hr
  • Response Time (Sec)9
  • Connector TypeSpade Lugs
  • Sheath Length (in)12
  • Sheath Diameter (InchShort)0.25
  • DescriptionSec. SPRT, 0.25" x 12" , -200~670C, Spade
  • Warranty1 year
Reviews
Q&A