The product you searched is discontinued. Below is the suggested replacement. Compare to discontinued item.
Cole-Parmer Ultrasonic Processor Continuous Medium Flow Cell
Representative image only.
Cole-Parmer Ultrasonic Processor Continuous Medium Flow Cell
Cole-Parmer | Item: 0471093
Recommended for the treatment of low-viscosity samples where the required insonation time is short
₹443,000.00INR / Each
₹443,000.00INR / Each
Product Description
Key Features
  • Made of autoclavable stainless steel
  • 3 year warranty
More About this Item
This low-volume cell screws onto the standard 2" dia probe and forces all of the sample solution to pass across the probe face. Use the cell to emulsify, disperse, and homogenize samples at flow rates up to 5 GPH (19 liters/hour). Fittings accept 16" ID soft tubing.
3 Year Warranty
Specifications
  • DescriptionUltrasonic Processor Continuous Medium Flow Cell