The product you searched is discontinued. Below is the suggested replacement. Compare to discontinued item.
Cole-Parmer Pipette Rack, 0.1 to 10 µL Tip; 10/PK
Representative image only.
Cole-Parmer Pipette Rack, 0.1 to 10 µL Tip; 10/PK
Cole-Parmer | Item: 2450304
Stackable color-coded trays to house your tips in a safe and contaminate-free environment
₹1,800.00INR / Pkg of 10
₹1,800.00INR / Pkg of 10
Product Description
Key Features
  • Fully autoclavable and stackable
  • Color-coded tray for easy identification
  • Hinge rack allows single hand operation
  • Free from DNase, RNase, Human DNA, Pyrogen and PCR inhibitors
  • Holds 0.1 to 10 µL tips
More About this Item
When it comes to reaching for that specific pipette tip, these racks make is easy. They can hold standard and long tips with various volume measurements and, are available in packages of 6 or 10 trays.
Specifications
  • Max Volume (µL)10
  • AutoclavableYes
  • Min Volume (µL)0.1
  • Tip StyleStandard
  • Color CodeYes
  • Package Of10 racks
  • Compatible withStandard 3.13 cm length tips
  • DescriptionOmega Pipette Racks for 0.1 to 10 µL tips, 10/PK