Cole-Parmer pH electrode-Flat, Sealed, 127 mm x 12 mm
Representative image only.
Cole-Parmer Item # WW-05990-65
Combines quality and durability
  • Epoxy-body housing resists impact
  • 6 month warranty
PRODUCT OPTIONS
₹20,600.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsLow volume sampling
  • Electrode TypeSingle junction
  • Electrode Body MaterialEpoxy
  • Reference ElectrolyteAg/AgCl
  • Max Temperature (° C)80
  • ConnectorBNC
  • DescriptionpH electrode-Flat, Sealed, 127 mm x 12 mm
  • Warranty6 month warranty
6 Month Warranty
Key Features
  • Epoxy-body housing resists impact
  • 6 month warranty
More About this Item
These quality laboratory electrodes have a sealed gel reference unless otherwise noted.