Oakton by Cole-Parmer® pH electrode, combination, spear tip, sealed, glass body, BNC
Representative image only.
Oakton Item # WW-05998-20
Withstand high temperatures and chemical attack
  • Exhibit minimal solution carryover
  • 6 month warranty
PRODUCT OPTIONS
₹14,800.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Electrode TypeSingle junction
  • Electrode Body MaterialGlass
  • Reference ElectrolyteAg/AgCl
  • Max Temperature (° C)100
  • ConnectorBNC
  • DescriptionpH electrode, combination, spear tip, sealed, glass body, BNC
  • Warranty6 month warranty
6 Month Warranty
Key Features
  • Exhibit minimal solution carryover
  • 6 month warranty
More About this Item
Glass-body combination electrodes feature refillable references, except for 05998-30 which features a high-temperature KNO3 gel. All have BNC connectors.