Environmental Express General purpose-lab, pH electrode, Refillable
Representative image only.
Environmental Express General purpose-lab, pH electrode, Refillable
Environmental Express | Item: 0599122
Faster response and increased stability across a pH range of 0 to 14
₹29,200.00INR / Each
₹29,200.00INR / Each
Buy It Together
Current Item
Environmental Express General purpose-lab, pH electrode, Refillable
Environmental Express General purpose-lab, pH electrode, Refillable
0599122
₹29,200.00INR / Each
+ +
Environmental Express Oakton Fill Solution 4.0M KCL 2/Pk
0599248
₹8,200.00INR / Pkg of 2
+ =
Total Price 68900.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Dome bulb offers better protection from breakage
  • Stability in low-ionic strength solutions
  • Offers resistance to Na+ ion interference
  • Compatible with most brands of pH meters (that accept standard BNC connections)
  • 6 month warranty
More About this Item
Glass body combination probes are able to withstand higher temperatures and chemical attack. All have 3.3 ft (1 m) long cables with BNC connectors.
6 Month Warranty
Specifications
  • ApplicationsGeneral purpose-lab
  • Electrode TypeSingle junction
  • Electrode Body MaterialGlass
  • Reference ElectrolyteAg/AgCl
  • Max Temperature (° C)100
  • ConnectorBNC
  • DescriptionGeneral purpose-lab, pH electrode, Refillable
  • Warranty6 month warranty