EchoTherm Programmable Aluminum Top Hot Plates
Store up to 10 programs in memory for instant recall and use
  • Hot plate heats up to maximum temperature in approximately 2 minutes
  • Platinum RTD temperature measurement for ±1% accuracy
Repeat any program up to 98 times automatically. Multiple temperatures, temperature ramp rates, and timed events can be stored. Includes timer to 99 hours, 59 minutes with auto-off option. Unit can be programmable or non-programmable in operation. Offers RS-232 interface for control or recording data via a computer.
344,000.00 - ₹524,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Top Plate Length (in)
Availability Pricing
Torrey Pines EchoTherm HP60A Hot Plate, Programmable Aluminum, 6" x 6", 115V 5180895 Mfr # HP60A
COMPARE
115 6
₹344,000.00INR / Each
Torrey Pines EchoTherm HP60A-2 Hot Plate, Programmable Aluminum, 6" x 6", 230V 5180897 Mfr # HP60A-2
COMPARE
230 6
₹344,000.00INR / Each
Torrey Pines EchoTherm HP61A Hot Plate, Programmable Aluminum, 12" x 12", 115V 5180903 Mfr # HP61A
COMPARE
115 12
₹524,000.00INR / Each
Torrey Pines EchoTherm HP61A-2 Hot Plate, Programmable Aluminum, 12" x 12", 230V 5180905 Mfr # HP61A-2
COMPARE
230 12
₹524,000.00INR / Each