Dissolved Oxygen (DO) Measurement FAQs
1. How should I store my DO probes?
Store a DO probe with the membrane covered by distilled water. Covering the membrane with distilled water prevents the KCl fill solution from evaporating through the membrane. 2. What is the difference between a galvanic and polarographic electrode?
A galvanic electrode produces its own current. The electrode is usually silver and lead with a potassium hydroxide electrolyte. 3. How does temperature affect my DO readings?
Temperature affects DO readings in two ways. First, it changes the permeability of the membrane. As the temperature increases so does the permeability of the membrane. As the temperature decreases the oxygen permeability through the membrane decreases. Secondly, temperature affects the amount of oxygen that can be dissolved in water. As temperature increases the oxygen saturation point of water decreases. Most meters have automatic temperature compensation (ATC) to compensate for the temperature error due to changes in the permeability of the membrane only. To compensate for saturation changes use a correction chart. There is a correction chart for fresh water and another for sea water. 4. How does atmospheric pressure affect my DO readings?
Oxygen saturation of water is different at different pressures. Correction charts are provided in most instruction manuals. Many instruments automatically compensate for barometric pressure. The barometric pressure is either measured by the instrument or entered by the user. 5. How do I calibrate my DO probe?
Zero calibrate the probe by submersing it in a saturated solution of sodium sulfite. For full-scale calibration recommends the forced air method. Place the probe in the middle of a vortex so that the air can be forced through the membrane quickly. Air can be forced through the membrane faster in a vortex than if the probe were simply sitting out in the air. A vortex can be created using a stirrer, stir bar, beaker of distilled water, and a probe holder to keep the probe above the water. 6. I just received my DO meter and it does not work, is there a serious problem?
No. This is common. The electrode must be allowed to polarize before any readings can be taken. In order to polarize the electrode must be connected to a meter. Polarization time varies from meter to meter so polarization could be from 10 minutes to 6 hours. Check your manual for the exact time. 7. What is BOD?
BOD is a measure of the amount of oxygen that is consumed by bacteria as they decompose the organic components of waste. DO measurements are usually made at the beginning and end of a five day incubation period for a standard test. 8. What is SOUR/DOUR?
9. How often should I replace my membranes?
Recommended service time is 2-4 weeks depending on the application. 10. My probe is not reading correctly, what could be the reasons and what can be done?
Try the following: |