Idex® PEEK Sample Loops for Injection Valve Models 3725 and 3725i
Biocompatible and inert to almost all organic solvents
  • Excellent alternative to stainless steel loops
  • Easy to install or replace
Biocompatible PEEK polymer sample loops are inert to almost all organic solvents, making them a versatile alternative to stainless steel. The loop ends are cut clean and straight for easy valve installation.
89,600.00 - ₹119,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Total Volume Capacity (mL) Tubing Inside Diameter (ID) (mm) Tubing Outside Diameter (OD) (in)
Availability Pricing
Idex Sample Loop for 3725/3725i Valves, PEEK, 5 mL, 1/8" OD x 0.062" ID; 1/EA 0201669 Mfr # 3055-019
COMPARE
5 1.6 0.125
Call For Price
Idex Sample Loop for 3725/3725i Valves, PEEK, 10 mL, 1/8" OD x 0.080" ID; 1/EA 0201670 Mfr # 3055-023
COMPARE
10 2 0.125
₹89,600.00INR / Each
Idex Sample Loop for 3725/3725i Valves, PEEK, 20 mL, 1/8" OD x 0.080" ID; 1/EA 0201671 Mfr # 3055-025
COMPARE
20 2 0.125
₹119,000.00INR / Each