EchoTherm Digital Hot Plates, Aluminum Top
Temperature ramping provides exact heating and cooling rates
  • PID digital control for precise plate and probe temperatures
  • Timer allows hands-free operation
Set temperature ramp rate from 1°C/hr to 400°C/hr. Platinum RTD sensor provides precise and accurate measurements over entire temperature range. Memory stored temperature calibration can be reset using the probe calibration kit (sold separately below). Set timer with auto-off for exact timed heating of your sample.
All models have an epoxy-coated cast aluminum chassis and a splash-resistant keyboard cover. Indicator lights let you know if top plate is hot or unit has reached overtemperature status. Aluminum top plates are ideal for general-purpose applications and can reach maximum temperature in under 10 minutes.
376,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC)
Availability Pricing
Torrey Pines EchoTherm HP30A-2-CP Digital Hot Plate, Aluminum, 230 VAC 5180815 Mfr # HP30A-2-CP
COMPARE
230
₹376,000.00INR / Each