Nilfisk Conductive Dust Brush for GM80CR Cleanroom Vacuum, 38 mm

Representative image only.

Nilfisk Conductive Dust Brush for GM80CR Cleanroom Vacuum, 38 mm

Nilfisk | Mfr: 147 1494 500| Item: 8656481

Enhance the performance of your vacuum

₹12,300.00INR / Each

₹12,300.00INR / Each
Product Description

Key Features

  • 2 year warranty

More About this Item

Keep your vacuum running at its best with these replacement parts and accessories.

Two year warranty
Specifications
  • ColorBlack
  • Compatible WithGM80CR
  • Diameter (InchShort)1 1/2
  • Diameter (cm)3.8
  • DescriptionConductive Dust Brush for GM80CR Cleanroom Vacuum, 38 mm
  • Warranty2 year
Reviews
Q&A