Cole-Parmer Filling Bell, Polycarbonate, 10/Pk
Representative image only.
Cole-Parmer Filling Bell, Polycarbonate, 10/Pk
Cole-Parmer | Item: 5014295
Safely transfer sterile media from large containers to smaller vessels
₹160,000.00INR / Pkg of 10
₹160,000.00INR / Pkg of 10
Product Description
Key Features
  • Clear polycarbonate for easy visibility while filling
  • Measures 11 cm H x 7.8 cm ID (4-11/32" x 3-1/16") with tubing adapter
  • 10 mm OD tube adapter
  • ASME-BPE, USP Class VI, FDA 21 CFR 177.1580, and RoHS compliant
  • Animal-derived ingredient free (ADIF) and animal-derived component free (ADCF)
  • Sterilize by autoclave, gamma irradiation, x-ray, or EtO
More About this Item
Connect tubing to the port at the top of the filling bell and place the filling bell over the receiving vessel for easy aseptic fluid transfer. The single-use filling bells protect your sterile media against airborne contaminants—ideal for use in the pharmaceutical, biotech, food, beverage, and other sanitary industries. The clear polycarbonate filling bells allow for visibility while filling and are lightweight for easy handling.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Height (InchShort)4 11/32
  • Regulatory ApprovalsFDA;RoHS;USP;ADCF;Non-Phthalate
  • DescriptionFilling Bell for Aseptic Fluid Transfer, Polycarbonate, 10/Pk