Monitor your cold chain accurately using temperature data loggers with probes
Traceable Excursion-Trac Data Logging Thermometer 1 bottle probe
By Ayisha Malik, EMEA
When recording temperature measurements of your vaccines’ or samples’ critical environments, a seemingly obvious but often overlooked point is that you are only ever recording the temperature of the sensor in your device. This can present direct errors, when there are discrepancies between the sample temperature and what is being captured by your temperature measurement device. To overcome this challenge, select a temperature instrument with a probe. A variety of Traceable® temperature instruments are equipped with probes. Some of the instruments have interchangeable probes, while others come with one specific probe. They are specifically designed to capture accurate temperature readings under different conditions. Here are the types of probes available:
Bottle probes
Bottle probes on a Traceable® Wi-Fi Data Logging Refrigerator/Freezer Thermometer
Bottle probes on a Traceable® Wi-Fi Data Logging Refrigerator/Freezer Thermometer.
Bottle probes are used to record the temperature of liquids stored in temperature-controlled environments. The 25-mL bottle is filled with a patented non-toxic glycol solution that is Generally Recognised as Safe (GRAS) by the Food and Drug Administration (FDA), eliminating concerns of incidental contact with food or drinking water. The solution-filled bottles simulate the temperature conditions of other stored liquids in the fridge or freezer, producing accurate readings for the liquids in that environment. Additionally, it eliminates fluctuations in the temperature readings can
used by opening and closing of fridge doors, which may be seen when using other types of probes.
Vaccine bottle probes
Vaccine-bottle probes on a Traceable® Excursion-Trac®  Refrigerator/Freezer Thermometer Data Logger
Vaccine-bottle probes on a Traceable® Excursion-Trac®  Refrigerator/Freezer Thermometer Data Logger.
Like the bottle probes, vaccine bottle probes are also filled with a non-toxic glycol solution. The 5-mL bottle mimics the liquid and volume of a standard vaccine sample to produce accurate temperature readings for vials in storage. The probes are also designed to fit perfectly into standard vaccine trays and simulate storage condition to accurately capture temperature changes.
Bullet probe on a TraceableGO® Temperature and Temperature/Humidity Bluetooth® Data Logger
Bullet probe on a TraceableGO® Temperature and Temperature/Humidity Bluetooth® Data Logger.
Bullet probes
The bullet probe is suitable for accurately recording temperatures of both liquids and gases. The sensor can be completely immersed in a fluid for precise temperature monitoring of both liquid products or liquid mediums that are often used to store and transport dangerous or flammable chemicals.
Stainless steel probes
Stainless steel probe on a Traceable® Ultra-Low Temperature Wi-Fi Data Logger
Stainless steel probe on a Traceable® Ultra-Low Temperature Wi-Fi Data Logger.
Stainless steel probes can withstand both cold and hot temperatures; and therefore, are ideal for capturing a broader temperature range compared to many of its counterparts. It is suitable for measuring temperatures of gases, liquids, and semi-solids, making it ideal for a wide array of products, samples, specimens and reagents.
Summary
Sectors, such as food and beverage, biopharma, healthcare, and academia, require careful temperature-controlled storage, transport and monitoring amenities. With an extensive portfolio of temperature measurement tools, offering a wide array of features, Traceable has an instrument to meet your needs. Whether you require one-off accurate readings, continuous temperature monitoring and reporting capabilities, or round-the-clock real-time updates from your critical environments, you can rely on Traceable for the right solution.
Need a reliable data logger? The Traceable Excursion-Trac Data Logging Refrigerator/Freezer Thermometers, shown as the featured image above, are ILAC-MRA accredited. Each instrument comes with its own, individually numbered NIST-traceable certificate and a choice of one or two bottle, vaccine bottle, bullet, or stainless steel probe(s).
COle-Parmer Blog
Related Articles
Why Calibiraion is Critical for Accurately Monitoring Vaccine Temperatures
Why Use USB Data Loggers in Clinical Trials
Considerations for Compliant Vaccine Temperature Monitoring During Flu Season
Learn more about Cold Chain: What is Cold Chain?
Be the first to comment on "Monitor your cold chain accurately using temperature data loggers with probes"