3 Ways to Use Environmental Chambers
Constant Climate Chamber
Constant Climate Chamber
Testing products in their actual environment provides significant feedback as to how they will work in real-world conditions. An environmental chamber delivers this feedback by exposing the material inside to stresses such as vibration, sun exposure, extreme temperatures, moisture, and more.
Will the product stand up to these conditions or will it disintegrate sooner than expected? The environmental test chamber can:
  • Help evaluate product quality and reliability, and identify manufacturing flaws and weaknesses
  • Show the process of product decay and degradation and help predict its potential lifespan
  • Assess how materials—including biological, industrial, and electronics—react to selected conditions
While there are various types of environmental chambers on the market, some include the constant climate chamber, temperature and humidity chamber, controlled environment chambers , mini humidity chambers, and climatic simulation and photostability test chambers. Consider all the ways you will need to test your product or material before making your selection.
Be the first to comment on "3 Ways to Use Environmental Chambers"