QRP VHC12 MEDIUM Vinyl Cleanroom Gloves, medium, 100/bag
Representative image only.
QRP VHC12 MEDIUM Vinyl Cleanroom Gloves, medium, 100/bag
QRP | Mfr: VHC12 MEDIUM| Item: 8658021
Perfect for pharmaceuticals, electronics, and cleanroom food processing
₹9,100.00INR / Bag of 100
₹9,100.00INR / Bag of 100
Product Description
Key Features
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
More About this Item
Gloves feature a micro-textured surface for enhanced gripping. Latex-free, 100% PVC co-polymer is naturally antistatic with a resistivity of 1010 ohms. Beaded cuff for additional strength. Ambidextrous. Packed 100 gloves per polybag.
Compliance: ISO Class 5, USDA, and IEST-RP-CC-005.2.
Specifications
  • MaterialVinyl
  • Thickness (mil)6
  • SizeMedium
  • Length (in)12
  • Length (cm)30.5
  • ColorWhite
  • GripMicro-textured
  • ComplianceISO Class 5; USDA; IEST-RP-CC-005.2
  • SterileNo
  • DescriptionVinyl Cleanroom Gloves, medium, 100/bag