Fluidic solutions for your vaccine and therapeutics production
Optimize your processes and reduce risk of contamination with our wide variety of fluidics products from Masterflex®, Ismatec®, and other trusted brands. Add intuitive and accurate peristaltic pumps and pump tubing, MasterflexLive® remote pump control and monitoring, flowmeters and controllers, single-use components, and bottle assemblies to keep your vaccine production flowing and valuable product safe to the finish line.
FERMENTATION
At the heart of your vaccine or therapeutics process, fermentation creates the proteins for your therapies. Careful monitoring and precise addition of media and other additives are essential for effective growth. Ensure optimal output with our wide range of fluid handling products.
-
Efficient, programmable feed and filling of media; flows to 19 LPM
-
Fluid transfer in critical process steps; easy EtherNet/IP automation integration; flows to 19 LPM
-
Programmable dosing of pH adjusters and other adjuncts; flows to 3400 mL/min
-
Accurate transfer and filling of media; flows to 3400 mL/min
-
Low-flow, programmable dosing of pH adjusters and other adjuncts; flows to 365 mL/min
HARVEST
From washing to buffer injection to filtering, the steps in harvesting begin to separate the protein from the cells. Look to our pumps for fluid transfer capabilities as well as single-use lines between filters and vessels. Depending on your production scale, our filtration products may also provide critical performance in your process.
-
Sanitary, low-pulsation pump delivers flow and pressure for filtration; flows to 3 LPM; pressure to 58 psi (4 bar)
-
Sanitary, shear-sensitive pump delivers flow and pressure for filtration; flows to 14 LPM; pressure to 58 psi (4 bar)
-
Cytoflow® pump is ideal for live cells, large molecules, and other shear-sensitive fluids; flows to 13.6 LPM
-
Cytoflow pump is ideal for pumping live cells, large molecules, and other shear sensitive fluids; flows to 5 LPM
-
Accurate, programmable sampling by volume or time; flows to 365 mL/min
PURIFICATION
Separate your final product from all remaining contaminants and effluvia. Concentrate and refine the proteins to prepare them for final product fill/finish. The isolated fluid flow path of peristaltic pumps is critical to this workflow.
-
Sanitary, low-pulsation pump delivers flow and pressure for filtration; flows to 3 LPM; pressure to 58 psi (4 bar)
-
Cytoflow® pump is ideal for live cells, large molecules, and other shear-sensitive fluids; flows to 13.6 LPM
-
Sanitary, shear-sensitive pump delivers flow and pressure for filtration; flows to 14 LPM; pressure to 58 psi (4 bar)
-
Accurate fluid transfer in critical process steps; flows to 19 LPM
-
Programmable sampling by volume or time; easy automation integration with EtherNet/IP; flows to 3400 mL/min
PROCESS ESSENTIALS
In addition to pumps and tubing to meet your process needs, select from a wide range of single-use assemblies and components, bottle assemblies, flow sensors, and pumps with remote control and monitoring to keep your vaccine production up and running around the clock without worry.
Get accurate flow readings without adding a new material to the fluid path; noninvasive design eliminates contact with working fluid
Interface between ultrasonic flow sensor and Masterflex pump receives input from flow sensor then adjusts pump speed to a set flow rate or dispense volume
Safely transfer, sample, and store biopharmaceutical products and reagents
Masterflex and Ismatec precision extruded and pump tested formulations engineered for the highest reliability, compliance, and traceability
Ensure secure, sanitary connections in critical process applications. Fittings and clamps in sizes needed from pilot to production