Zymit Pro Enzyme Cleaner
Biodegradable enzyme and surfactant cleaner that breaks down and removes proteinaceous soils.
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
It is an effective filter membrane cleaner, especially when used with Micro-90 or Micro A07 in consecutive cleaning steps.
48,800.00 - ₹530,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Dilution Format Description
Availability Pricing
International Products Corp Z-0701-12 Zymit Pro Enzyme Cleaner, 12 x 1 L WW-18100-17 Mfr # Z-0701-12
COMPARE
1:100 to 2:100, pH 7.5 Liquid Z-0701-12 ZYMIT PRO ENZYME CLEANER 21.0 KG NET WT CONTAINER
₹48,800.00INR / Case of 12
International Products Corp Z-0720 Zymit Pro Enzyme Cleaner, 21 KG (19 L) WW-18100-18 Mfr # Z-0720
COMPARE
1:100 to 2:100, pH 7.5 Liquid Z-0720 ZYMIT PRO ENZYME CLEANER 21.0 KG NET WT CONTAINER
₹64,300.00INR / Each
International Products Corp Z-0790 Zymit Pro Enzyme Cleaner, 215 KG (197 L) WW-18100-19 Mfr # Z-0790
COMPARE
1:100 to 2:100, pH 7.5 Liquid Z-0790 ZYMIT PRO ENZYME CLEANER 215 KG NET WT PLASTIC DRUM
₹530,000.00INR / Each