This product is now discontinued, View Related Products
ZoneDispo Disposable Solids Samplers
Sample three zones at once
  • Three spaced openings along shaft sample different layers in one motion
  • Free-flowing powders and small-grain granulates work best
  • Rotating handle closes sampler for transport
This HDPE powder and granulate sampler provides a fast all-in-one solution to layer sampling. With three openings spaced apart along the hollow shaft, the product falls into the sampler from three distinct locations simultaneously. No need to repeat the sampling process multiple times. The handle is closed and when rotated, the openings are closed off providing a secure container for transport.
Gamma sterilized and nonsterile packs available.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Length (in) Description
Availability Pricing
Dynalon ZoneDispo Three-Zone Solids Sampler, 100 mL, 500 mm L; 20/pk WW-33687-16 Mfr # 853933-4420
COMPARE
19.625 Three-Zone Solids Sampler, 100 mL, 500 mm L; 20/pk
Call For Price
Dynalon ZoneDispo Sterile Three-Zone Solids Sampler, 100 mL, 500 mm L; 20/pk WW-33687-17 Mfr # 853933-4421
COMPARE
19.625 Sterile Three-Zone Solids Sampler, 100 mL, 500 mm L; 20/pk
Call For Price
Dynalon ZoneDispo Three-Zone Solids Sampler, 200 mL, 1 m L; 20/pk WW-33687-18 Mfr # 853933-4430
COMPARE
39.375 Three-Zone Solids Sampler, 200 mL, 1 m L; 20/pk
Call For Price
Dynalon ZoneDispo Sterile Three-Zone Solids Sampler, 200 mL, 1 m L; 20/pk WW-33687-19 Mfr # 853933-4431
COMPARE
39.375 Sterile Three-Zone Solids Sampler, 200 mL, 1 m L; 20/pk