YSI Replacement Membrane Kits
Available in standard or high-sensitivity versions
  • Each kit contains 30 membranes and 5.25 g of KCL crystals.
Use the standard kit (05522-00) for general laboratory DO needs; or choose the high-sensitivity kit (05522-10) for oxygen levels <= 5 mg/L at 15°C or below.
11,200.00 - ₹20,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
YSI 5775 Standard Membrane Kit for 05521-00; 30 Membranes, 5.25g KCl WW-05522-00 Mfr # 098094
COMPARE
Standard Membrane Kit for 05521-00; 30 Membranes, 5.25g KCl
₹11,200.00INR / Each
YSI 5776 High-Sensitivity DO Membrane Kit WW-05522-10 Mfr # 098095
COMPARE
High-Sensitivity DO Membrane Kit
₹20,000.00INR / Each