YSI ProOBOD probe
Optical DO technology reduces maintenance
  • Smart sensor retains calibration—no need to recalibrate when switching instruments
  • Fits standard 300 mL US/Japan or International BOD bottles
  • Self-stirring probe ensures a representative sample and improves response time
YSI ProOBOD probe uses optical measurement technology for BOD applications. Optical technology provides greater measurement stability, and reduces probe maintenance and calibration costs for long-term savings.
Auto-stirring mechanism provides faster response time and ensures a representative sample. OBOD probe has a keyed military spec connector design allowing the OBOD to be easily removed from the meter. Rubber overmolding allows for comfortable handling from any angle.
273,000.00 - ₹277,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
YSI 626400 ProOBOD Probe, US/Japanese version, self-stirring WW-59353-30 Mfr # 626400
COMPARE
ProOBOD Probe, US/Japanese version, self-stirring
₹273,000.00INR / Each
YSI 626401 ProOBOD probe, international version WW-59353-31 Mfr # 626401
COMPARE
ProOBOD probe, international version
₹277,000.00INR / Each