This product is now discontinued, View Related Products
Yamato Vertical-Loading Portable Sterilizers
Vertical loading maximizes usable chamber space
  • Casters and flexible plumbing make sterilizers completely mobile
  • Water-resistant keypad protects against steam and condensation
  • Front door water level site glass shows you when the water tank needs to be emptied
These compact portable sterilizers are ideal for laboratories that share equipment. Single-cycle models sterilize while dual-cycle models feature both sterilizing and drying cycles. Designed with rolling casters for excellent mobility and easy storage—just push it into a storage room when not in use to make room in your lab. Top mounted stainless steel door also saves space in your lab and provides easy loading.
The sterilizers feature fully automated operation. Simply place your load in the pressure chamber and enter all parameters with the easy-to-use keypad. The LED display shows set and actual temperature, as well as set time and time remaining.
Self-diagnostic circuitry monitors the temperature sensor, low-water sensor, and heater; alerts you with an audible and a visual alarm if a malfunction occurs. Additional safety features include a circuit breaker, safety pressure release valve, and a water level site glass.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Chamber Capacity (cu ft) Power (VAC)
Availability Pricing
Yamato SM-200 Vertical-Loading Portable Sterilizer; capacity, 0.8 cu ft; chamber WW-10770-02 Mfr # SM-200
COMPARE
0.8 115
Call For Price
Yamato SM-210 Vertical-Loading Portable Sterilizer; 0.8 cu ft, 220V WW-10770-07 Mfr # 10770-05 (reactivate)
COMPARE
0.8 220
Call For Price
Yamato SM-300 Vertical-Loading Portable Sterilizer; 1.2 cu ft, 115V WW-10771-02 Mfr # SM-300
COMPARE
1.2 115
Call For Price
Yamato SM-310 Vertical-Loading Portable Sterilizer; 1.2 cu ft, 220V WW-10771-07 Mfr # SM-310
COMPARE
1.2 220
Call For Price
Yamato SM-500 Vertical-Loading Portable Sterilizer; 1.8 cu ft, 115V WW-10772-02 Mfr # SM-500
COMPARE
1.8 115
Call For Price
Yamato SM-510 Vertical-Loading Portable Sterilizer; 1.8 cu ft, 220V WW-10772-07 Mfr # SM-510
COMPARE
1.8 220