This product is now discontinued, View Related Products

WTW pH Buffer Ampoules

Up to 5 years shelf life
  • Convenient ready-to-use, sealed glass ampoules with presterilized buffers
  • Certified accuracy traceable to NIST/DIN standards
Highly precise standards with +/-0.01 pH accuracy. Presterilized and free of preservatives, these single-use buffers offer a 5-year shelf life. Each sealed glass ampoule contains 25 mL of solution.
Call For Price

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Buffer Kit Values Volume (mL) Description
Availability Pricing
WTW 285138398 pH 4 & 7 Buffer Ampoules, 2 x 30Ea WW-59351-36 Mfr # 285138398
COMPARE
4 and 7 25 pH 4 & 7 Buffer Ampoules, 2 x 30Ea
Call For Price
WTW 285138632 4 & 7 Buffer Ampoules, 2x9 Ea, 50 mL 3M KCl WW-59351-38 Mfr # 285138632
COMPARE
4 and 7 25 4 & 7 Buffer Ampoules, 2x9 Ea, 50 mL 3M KCl
Call For Price
WTW 285138192 4,7,10 Buffer Ampoules, 2 x 30 Ampoules Ea WW-59351-42 Mfr # 285138192
COMPARE
4, 7, and 10 25 4,7,10 Buffer Ampoules, 2 x 30 Ampoules Ea
Call For Price
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat