WTW Conductivity Standard Ampoules
Offering up to 5 years shelf life
  • Highly precise standards with ±0.25% accuracy
  • Certified accuracy traceable to NIST/DIN standards
  • Single-use buffers with 25-mL of solution
Presterilized and free of preservatives, these ready-to-use, sealed glass ampoules of conductivity standards offer a 5-year shelf life. 25 mL solution per ampoule.
23,600.00INR / Pkg of 18
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
WTW 285126293 Conductivity Standard, 1.41/12.9/112 mS; 3 x 6 Ampoules Ea WW-59351-46 Mfr # 285126293
COMPARE
Conductivity Standard, 1.41/12.9/112 mS; 3 x 6 Ampoules Ea
₹23,600.00INR / Pkg of 18