Worthington (aka Taylor-Wharton) Dewars with Canister Storage Systems
Safely store samples up to eleven months without replenishing Liquid Nitrogen
These Dewars with canister and rack storage systems offer long-term, cost-effective storage for cells, tissues, and other biological samples. Canisters hold samples in either the vapor or liquid phase depending on the liquid nitrogen level. Temperatures are maintained from –184°C under the closed necktube core to –196°C at the liquid nitrogen level.
Choose from seven different capacities and three different neck diameters to suit your storage requirements. Dewar models 03778-36, -41 and 03779-31 require less frequent liquid nitrogen refilling for long-term, worry-free storage.
All models feature lightweight construction for easy handling. Ribbed-body design and high-strength aluminum construction ensure durability.
Each Dewar comes with canisters or racks to hold any combination of cryogenic vials, tubes, or sample containers. Canister handles are numbered for easy identification and trouble-free sample retrieval. To transport your Dewars easily, order an optional roller base below.
Use measuring rod 03778-60 (sold separately) to determine and maintain correct liquid nitrogen level.
230,000.00 - ₹918,000.00INR / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Neck Inside Diameter (ID) (in) Volume (Liters)
Availability Pricing
Worthington 20XTB-11M Dewars with Canister Storage Systems, 20.7 Liters, 2.0" Neck ID WW-03778-36 Mfr # 20XTB-11M
COMPARE
2 20.7
₹230,000.00INR / Each
Worthington 34XTB-11M Dewars with Canister Storage Systems, 34 Liters, 2.0" Neck ID WW-03778-41 Mfr # 34XTB-11M
COMPARE
2 34
₹276,000.00INR / Each
Worthington 20HCB-11M Dewars with Canister Storage Systems, 20 Liters, 3.6" Neck ID WW-03779-12 Mfr # 20HCB-11M
COMPARE
3 5/8 20
₹278,000.00INR / Each
Worthington 34HCB-11M Dewars with Canister Storage Systems, 34 Liters, 3.6" Neck ID WW-03779-31 Mfr # 34HCB-11M
COMPARE
3 19/32 34
₹281,000.00INR / Each
Worthington 35HCB-11M Cryogenic Storage Dewar System, 35 L; 10 Canisters WW-03779-41 Mfr # 35HCB-11M
COMPARE
4 11/16 35
₹350,000.00INR / Each
Worthington 35VHCB-11M Cryogenic Storage Dewar System, 35 L; 6 Canisters WW-03779-51 Mfr # 35VHCB-11M
COMPARE
4 11/16 35
₹361,000.00INR / Each
Worthington LS750B-R Dewars with Rack Storage Systems, 35 Liters, 4.0" Neck ID WW-03779-60 Mfr # LS750B-R
COMPARE
4 11/16 35
₹492,000.00INR / Each
Worthington LS3000 Dewars with Canister Storage Systems, 81 Liters, 8.5" Neck ID WW-03779-62 Mfr # LS3000
COMPARE
8 1/2 81
₹807,000.00INR / Each
Worthington LS4800 Dewars with Canister Storage Systems, 130 Liters, 8.5" Neck ID WW-03779-64 Mfr # LS4800
COMPARE
8 1/2 130
₹918,000.00INR / Each
Worthington LS6000 Dewars with Canister Storage Systems, 165 Liters, 8.5" Neck ID WW-03779-66 Mfr # LS6000
COMPARE
8 1/2 165
Call For Price
Showing 1- 10 of 10