This product is now discontinued, View Related Products
WorkBeads™ Affinity Chromatography Purification Media
Separate proteins using reverse interaction between the target protein and ligand of bead
  • Reliable, reproducible, and efficient
  • High chemical stability for easy clean-in-place (CIP)
Use affinity chromatography to separate biochemical mixtures based on a highly specific interaction between antigen and antibody, enzyme and substrate, or receptor and ligand.
Activated media are a reliable starting material for the preparation of customized chromatography resins. Activated media have a bromohydrin group that is reactive towards primary amines, thiol, hydroxyl, or histidyl residues. Select from two pore sizes depending on your application needs. Use the higher porosity media if the ligand or target is large.
Order media for antibodies for purification of monoclonal and polyclonal antibodies. The specificity of the recombinant Protein A for the Fc-region of IgG provides excellent purification. The Protein A ligand is coupled to the bead using a bromohydrin-based method that gives high chemical stability and low ligand leakage.
98,700.00 - ₹477,000.00INR / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Wetted Materials Media Volume (mL)
Availability Pricing
Bio-Works WorkBeads 40/1000 ACT Activated Medium, Bromohydrin, 1M kDa; 50 mL WW-11947-02 Mfr # KN40400001
COMPARE
Activated media 50
₹118,000.00INR / Each
Bio-Works WorkBeads 40/1000 ACT Activated Medium, Bromohydrin, 1M kDa; 300 mL WW-11947-03 Mfr # KN40400003
COMPARE
Activated media 300
₹435,000.00INR / Each
Bio-Works WorkBeads 40/1000 ACT Activated Medium, Bromohydrin, 1M kDa; 1 L WW-11947-04 Mfr # KN40400010
COMPARE
Activated media 1000
Call For Price
Bio-Works WorkBeads 40/10 000 ACT Activated Medium, Bromohydrin, 10M kDa; 50 mL WW-11947-05 Mfr # KN40450001
COMPARE
Activated media 50
₹118,000.00INR / Each
Bio-Works WorkBeads 40/10 000 ACT Activated Medium, Bromohydrin, 10M kDa; 300 mL WW-11947-06 Mfr # KN40450003
COMPARE
Activated media 300
₹437,000.00INR / Each
Bio-Works WorkBeads 40/10 000 ACT Activated Medium, Bromohydrin, 10M kDa; 1 L WW-11947-07 Mfr # KN40450010
COMPARE
Activated media 1000
Call For Price
Bio-Works WorkBeads 40 Protein A Affinity Medium for Antibodies (IgG); 1.5 mL WW-11947-17 Mfr # KN40605001
COMPARE
Affinity media 1.5
Call For Price
Bio-Works WorkBeads 40 Protein A Affinity Medium for Antibodies (IgG); 5 mL WW-11947-18 Mfr # KN40605002
COMPARE
Affinity media 5
Call For Price
Bio-Works WorkBeads 40 Protein A Affinity Medium for Antibodies (IgG); 10 mL WW-11947-19 Mfr # KN40605003
COMPARE
Affinity media 10
₹98,700.00INR / Each
Bio-Works WorkBeads 40 Protein A Affinity Medium for Antibodies (IgG); 100 mL WW-11947-20 Mfr # KN40605004
COMPARE
Affinity media 100
₹477,000.00INR / Each
Bio-Works WorkBeads 40 Protein A Affinity Medium for Antibodies (IgG); 1000 mL WW-11947-21 Mfr # KN40605005
COMPARE
Affinity media 1000
Call For Price
Showing 1- 11 of 11