Porta-Lab Wire Gauze for use with Bunsen Burners
Gauze is made of iron wire (24 SWG/10 mesh) for long life and safety
  • Withstands temperatures up to 1472°F (800°C)
  • Edges are reinforced to prevent fraying.
4,300.00 - ₹6,100.00INR / Pkg of 10
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Length (in) Width (in) Description
Availability Pricing
Porta-Lab 2390-4X4 Laboratory Wire Gauze, Square; 4" L x 4" W (10 Per Pk) WW-36300-40 Mfr # WGP44
COMPARE
4 4 Laboratory Wire Gauze, Square; 4\" L x 4\" W (10 Per Pk)
₹4,300.00INR / Pkg of 10
Porta-Lab 2390-5X5 Laboratory Wire Gauze, Square; 5" L x 5" W (10 Per Pk) WW-36300-42 Mfr # WGP55
COMPARE
5 5 Laboratory Wire Gauze, Square; 5\" L x 5\" W (10 Per Pk)
₹5,100.00INR / Pkg of 10
Porta-Lab 2390-6X6 Laboratory Wire Gauze, Square, 6 " L x 6" W (10 Per Pk) WW-36300-44 Mfr # WGP66
COMPARE
6 6 Laboratory Wire Gauze, Square, 6 \" L x 6\" W (10 Per Pk)
₹6,100.00INR / Pkg of 10