This product is now discontinued, View Related Products
O C White Wide-View Dimmable Illuminated Magnifiers
Dim the light intensity from 100 to 25% to reduce eye strain
  • Equipped with an ESD-safe shade
  • Full-spectrum bulbs for high-quality light output
Slide switch allows you to vary the light output from 100 to 25% with the stroke of a finger. The "U"-shaped bulb provides three-sided illumination — eliminates shadows that can occur with two bulbs. Large 7-1/2" x 6-3/16" (19.1 x 15.7 cm) lens gives an expanded viewing area. Shade handles and swivel mount allow easy positioning — rotating locking handles lock in your position. Magnifiers operate on 120 VAC.
129,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Arm Length (in) Diopters Magnification
Availability Pricing
O C White 72400 Wide-View Dimmable Illuminated Magnifier, 43" reach, 3 diopters WW-41752-20 Mfr # 72400
COMPARE
43 3D 1.85x
₹129,000.00INR / Each
O C White 72400-4 Wide-View Dimmable Illuminated Magnifier, 43" reach, 4 diopters WW-41752-25 Mfr # 72400-4
COMPARE
43 4D 2x
Call For Price
O C White 71400 Wide-View Dimmable Illuminated Magnifier, 25" reach, 3 diopters, table clamp WW-41752-30 Mfr # 71400
COMPARE
25 3D 1.85x
Call For Price
O C White 71600 Wide-View Dimmable Illuminated Magnifier, 25" reach, 3 diopters, weighted base WW-41752-31 Mfr # 71600
COMPARE
25 3D 1.75x to 1.85x