WHEATON® Glass Specimen Vials
Reliably collect and store samples
  • Manufactured from WHEATON® 800 soda-lime clear glass that conforms to USP Type III requirements
Ideal for economical collection and storage of all types of samples. Clear soda-lime glass is chemically durable.
LDPE snap-on caps form an air tight seal perfect for moisture sensitive materials. Finger tab on cap allows easy removal.
Vials are packed in convenient pull-out trays, two trays per case. Case protects vials during transport and is ideal for storage.
23,900.00 - ₹37,900.00INR / Case of 144
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cap Diameter (mm) Tube Material Volume (mL)
Availability Pricing
DWK Life Sciences (Wheaton) 225534 Glass Specimen Vials, Clear with LDPE Snap Caps, 8 mL; 144/Cs WW-08921-01 Mfr # 225534
COMPARE
22 Soda-Lime Glass 8
₹30,000.00INR / Case of 144
DWK Life Sciences (Wheaton) 225535 Glass Specimen Vials, Clear with LDPE Snap Caps, 12 mL; 144/Cs WW-08921-02 Mfr # 225535
COMPARE
22 Soda-Lime Glass 12
₹37,700.00INR / Case of 144
DWK Life Sciences (Wheaton) 225536 Glass Specimen Vials, Clear with LDPE Snap Caps, 16 mL; 144/Cs WW-08921-03 Mfr # 225536
COMPARE
30 Soda-Lime Glass 16
₹37,900.00INR / Case of 144
DWK Life Sciences (Wheaton) 225543 Glass Specimen Vials, Clear with LDPE Snap Caps, 30 mL; 72/Cs WW-08921-05 Mfr # 225543
COMPARE
34 Soda-Lime Glass 30
₹23,900.00INR / Case of 72
DWK Life Sciences (Wheaton) 225546 Glass Specimen Vials, Clear with LDPE Snap Caps, 120 mL; 72/Cs WW-08921-07 Mfr # 225546
COMPARE
45 Soda-Lime Glass 120
₹30,900.00INR / Case of 72