Wheaton Safety-Coated Narrow-Mouth Bottles
Plastisol coating resists breakage and ensures a slip-resistant grip
Bottles are manufactured with glass that meets USP Type III standards; the clear plastisol coating resists acid and solvent degradation. All bottles include PTFE-lined caps.
Bottles are not autoclavable.
41,100.00 - ₹57,500.00INR / Case of 24
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap / Stopper Size Material
Availability Pricing
DWK Life Sciences (Wheaton) 220775 Safety-Coated Bottles with PTFE-Lined Caps; 16 oz, Clear, 24/cs WW-08922-50 Mfr # 220775
COMPARE
500 28-400 mm Glass
₹57,500.00INR / Case of 24
DWK Life Sciences (Wheaton) 220975 Safety-Coated Bottles with PTFE-Lined Caps; 16 oz, Amber, 24/cs WW-08922-55 Mfr # 220975
COMPARE
500 28-400 mm Glass
₹57,500.00INR / Case of 24
DWK Life Sciences (Wheaton) 220776 Safety-Coated Bottles with PTFE-Lined Caps; 32 oz, Clear, 12/cs WW-08922-60 Mfr # 220776
COMPARE
1000 33-400 Glass
₹41,200.00INR / Case of 12
DWK Life Sciences (Wheaton) 220976 Safety-Coated Bottles with PTFE-Lined Caps; 32 oz, Amber, 12/cs WW-08922-65 Mfr # 220976
COMPARE
1000 33-400 Glass
₹41,100.00INR / Case of 12