WHEATON® Liquid Scintillation Vials, PET
Reliably collect and store samples
  • Constructed of durable Polyethylene Terephthalate (PET)
These 20-mL scintillation vials are made of crystal clear polyethylene terephthalate (PET). Vials feature background counts are consistent and low and ultraviolet transmission is high, making them ideal for solvent reduction. Multiple vial and cap configuration options are available for a wide range of applications. Vials are available with cap attached or packed separately.
37,900.00 - ₹69,500.00INR / Pkg of 500
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Septum Type Special Features Volume (mL)
Availability Pricing
DWK Life Sciences (Wheaton) Liquid Scintillation Vials, PET, Polypropylene Cap with Foamed Polyethylene Liner, 22-400 Thread, 20 mL; 500/Pk WW-08918-83 Mfr # 986730
COMPARE
Foamed Polyethylene Caps are supplied attached to vials 20
₹38,000.00INR / Pkg of 500
DWK Life Sciences (Wheaton) Liquid Scintillation Vials, PET, Polypropylene Cap with Metal Foil/Pulp Liner, 22-400 Thread, 20 mL; 500/Pk WW-08918-85 Mfr # 986731
COMPARE
Metal Foil / Pulp Caps are supplied attached to vials 20
₹37,900.00INR / Pkg of 500
DWK Life Sciences (Wheaton) Liquid Scintillation Vials, PET, Polypropylene Cap with Foamed Polyethylene Liner, 22-400 Thread, 20 mL; 1000/Pk WW-08918-87 Mfr # 986740
COMPARE
Foamed Polyethylene Caps are packaged separately from vials 20
₹62,600.00INR / Pkg of 1000
DWK Life Sciences (Wheaton) Liquid Scintillation Vials, PET, Polypropylene Cap with Metal Foil/Pulp Liner, 22-400 Thread, 20 mL; 1000/Pk WW-08918-89 Mfr # 986741
COMPARE
Metal Foil / Pulp Caps are packaged separately from vials 20
₹69,500.00INR / Pkg of 1000