Wheaton® LVI Vials, Borosilicate Glass, 0.3 mL
Compatible with all autosamplers that use 12 x 32 mm vials
  • Available in clear or amber glass with choice of screw or crimp top
Large opening vials with sealed-in limited volume inserts. The glass inserts are fused to the vial which helps prevent autosampler needle damage and allows a more consistant sample recovery. Each vial is made from borosilicate galss and feature a white ceramic "write-on" patch for easy sample identification. Available with a either an 11 mm crimp top or a 9mm screw thread.
36,100.00 - ₹42,700.00INR / Case of 100
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cap Diameter (mm) Tube Material Volume (mL)
Availability Pricing
DWK Life Sciences (Wheaton) LVI 0.3 mL Clear Vial with 11mm Crimp Seal, PTFE/Rub, 100/CS WW-08917-05 Mfr # W225223-01
COMPARE
11 Borosilicate Glass 0.3
₹36,100.00INR / Case of 100
DWK Life Sciences (Wheaton) LVI 0.3 mL Clear Vial with 9mm Blue Screw Cap, PTFE/Sil, 100/CS WW-08917-07 Mfr # W225326-0204
COMPARE
9 Borosilicate Glass 0.3
₹40,800.00INR / Case of 100
DWK Life Sciences (Wheaton) LVI 0.3 mL Amber Vial with 9mm Blue Screw Cap, PTFE/Sil, 100/CS WW-08917-08 Mfr # W225328-0204
COMPARE
9 Borosilicate Glass 0.3
₹42,700.00INR / Case of 100