WHEATON® Liquid Scintillation Vials, HDPE
Reliably collect and store samples
  • Constructed of durable High Density Polyethylene (HDPE)
These 20-mL scintillation vials are made of lightweight and durable high density polyethylene (HDPE). Vials feature background counts are consistent and low and ultraviolet transmission is high, making them ideal for solvent reduction. Multiple vial and cap configuration options are available for a wide range of applications. Vials are available with cap attached or packed separately.
29,200.00 - ₹55,100.00INR / Pkg of 500
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Cap Material Septum Type Special Features
Availability Pricing
DWK Life Sciences (Wheaton) Liquid Scintillation Vials, HDPE, Urea Cap with Metal Foil / Cork Liner, 22-400 Thread, 20 mL; 500/Pk WW-08918-51 Mfr # 986702
COMPARE
Urea Metal Foil / Cork Caps are supplied attached to vials
₹37,200.00INR / Pkg of 500
DWK Life Sciences (Wheaton) Liquid Scintillation Vials, HDPE, Polyethylene Cap with No Liner, 22-400 Thread, 20 mL; 500/Pk WW-08918-52 Mfr # 986704
COMPARE
Polyethylene None Caps are supplied attached to vials
₹33,000.00INR / Pkg of 500
DWK Life Sciences (Wheaton) Liquid Scintillation Vials, HDPE, Polypropylene Cap with Metal Foil / Cork Liner, 22-400 Thread, 20 mL; 1000/Pk WW-08918-53 Mfr # 986721
COMPARE
Polypropylene Metal Foil / Cork Caps are packaged separately from vials
₹45,200.00INR / Pkg of 1000
DWK Life Sciences (Wheaton) Liquid Scintillation Vials, HDPE, Polypropylene Cap with Metal Foil / Cork Liner, 22-400 Thread, 20 mL; 500/Pk WW-08918-54 Mfr # 986701
COMPARE
Polypropylene Metal Foil / Cork Caps are supplied attached to vials
₹29,200.00INR / Pkg of 500
DWK Life Sciences (Wheaton) Liquid Scintillation Vials, HDPE, Polyethylene Cap with No Liner, 22-400 Thread, 20 mL; 1000/Pk WW-08918-62 Mfr # 986724
COMPARE
Polyethylene None Caps are packaged separately from vials
₹55,100.00INR / Pkg of 1000