WHEATON® LAB FILE® Glass Sample Vials
Lab File® with partitioned trays provides an easy way to inventory samples or to store empty vials
  • PTFE/14B rubber-lined black phenolic screw caps are ideal for organic samples and improved chemical compatibility
Large Borosilicate Vials are autoclavable and come in 20-, 25-, and 40-mL sizes with white rubber-lined screw cap. Packed in cardboard trays, 72 per case.
17,000.00 - ₹19,800.00INR / Case of 72
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Description Tube Material Volume (mL)
Availability Pricing
DWK Life Sciences (Wheaton) 225288 LAB FILE® Sample Vials, Glass with Rubber-Lined Cap, 20 mL; 72/Cs WW-08918-22 Mfr # 225288
COMPARE
Borosilicate Vials, Clear w/Rubber-Lined Cap, 20 mL; 72/Cs Borosilicate Glass 20
₹17,000.00INR / Case of 72
DWK Life Sciences (Wheaton) 225289 LAB FILE® Sample Vials, Glass with Rubber-Lined Cap, 25 mL; 72/Cs WW-08918-23 Mfr # 225289
COMPARE
Borosilicate Vials, Clear w/Rubber-Lined Cap, 25 mL; 72/Cs Borosilicate Glass 25
₹18,100.00INR / Case of 72
DWK Life Sciences (Wheaton) 225290 LAB FILE® Sample Vials, Glass with Rubber-Lined Cap, 40 mL; 72/Cs WW-08918-24 Mfr # 225290
COMPARE
Borosilicate Vials, Clear w/Rubber-Lined Cap, 40 mL; 72/Cs Borosilicate Glass 40
₹19,800.00INR / Case of 72