Wheaton® Headspace Vial Convenience Packs
Each convenience pack includes 100 vials and 100 pressure release aluminum seals with molded septa
  • Choice of two vial sizes and three liner options
Wheaton Headspace Vials feature rounded shoulders and bottoms, allowing for even heating and safer operation at higher pressures. The newly designed top provides a tighter seal with your septa. Vials are manufactured from Type I borosilicate glass.
26,600.00 - ₹28,300.00INR / Case of 100
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Height (mm) Volume (mL)
Availability Pricing
DWK Life Sciences (Wheaton) Headspace Vial, 10 mL, 20mm Al Seal, PTFE/Silicone Septa; 100/Cs WW-08917-21 Mfr # W225278-01
COMPARE
20 10
₹26,600.00INR / Case of 100
DWK Life Sciences (Wheaton) Headspace Vial, 20 mL, 20mm Al Seal, PTFE/Silicone Septa; 100/Cs WW-08917-31 Mfr # W225280-01
COMPARE
20 20
₹28,300.00INR / Case of 100