Wheaton® E-Z VIAL® Autosampler Crimp Top Vial Convenience Kits
Save time during sample preparation and reduce the risk of contamination
  • Vial kits consist of vials with E-Z Seal™ crimp top and seals packaged in a convenient tray
Borosilicate glass vials feature 40% larger openings than standard vials—providing improved sample access and minimizing needle damage. Supplied in easy-order convenience kits—including 100 vials and 100 pre-assembled seals with septa. For microvolume sampling, limited-volume inserts are available separately. Inserts feature bottom springs to cushion needle impact.
6,600.00 - ₹7,300.00INR / Pkg of 100
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cap Material Septum Type Volume (mL)
Availability Pricing
DWK Life Sciences (Wheaton) EZ Vial Convenience Kits with Clear Vial and PTFE/Red Rubber Septa WW-98760-01 Mfr # W225175-01
COMPARE
Aluminum PTFE/red rubber 1.8
₹6,600.00INR / Pkg of 100
DWK Life Sciences (Wheaton) Autosampler Vial Kit, 1.8 mL, Amber, Write-on, PTFE/Rubber; 100/Cs WW-98763-01 Mfr # W225173-01
COMPARE
Aluminum PTFE/red rubber 1.8
₹7,300.00INR / Pkg of 100