WHEATON® CryoELITE® Cryogenic Vials, Sterile, Polypropylene
Leakproof even during changing temperatures—ultimate protection during transport
  • Made from low binding, cryogenic-grade virgin polypropylene
  • Lot-certified vials are free of RNase/DNase and Endotoxin—ensure product integrity
Tubes and closures have same coefficient of expansion to enhance leakproof qualities. White marking area aids in easy sample identification. Vials withstand temperatures from 121°C to 196°C (250 to 384°F). Flat-bottom vials have conical shaped wells and are self standing. Exacting round-bottom vials allow centrifugation up to 17,000 max RCF (xG). Order racks separately. Autoclavable.
42,300.00 - ₹63,400.00INR / Case of 500
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cap color Thread Volume (mL)
Availability Pricing
DWK Life Sciences (Wheaton) CryoELITE® Cryogenic Vials, Sterile, Polypropylene, External Thread, Round Bottom, Natural Cap, 2 mL; 500/Cs WW-03756-06 Mfr # W985861
COMPARE
Natural External 2
₹46,100.00INR / Case of 500
DWK Life Sciences (Wheaton) CryoELITE® Cryogenic Vials, Sterile, Polypropylene, External Thread, Flat Bottom, White Cap, 2 mL; 500/Cs WW-03756-08 Mfr # W985863
COMPARE
White External 2
₹50,100.00INR / Case of 500
DWK Life Sciences (Wheaton) CryoELITE® Cryogenic Vials, Sterile, Polypropylene, External Thread, Flat Bottom, Yellow Cap, 2 mL; 500/Cs WW-03756-14 Mfr # W985866
COMPARE
Yellow External 2
₹50,100.00INR / Case of 500
DWK Life Sciences (Wheaton) CryoELITE® Cryogenic Vials, Sterile, Polypropylene, External Thread, Flat Bottom, Green Cap, 2 mL; 500/Cs WW-03756-16 Mfr # W985867
COMPARE
Green External 2
₹50,100.00INR / Case of 500
DWK Life Sciences (Wheaton) CryoELITE® Cryogenic Vials, Sterile, Polypropylene, Internal Thread, Flat Bottom, Natural Cap, 1.2 mL; 500/Cs WW-03756-24 Mfr # W985915
COMPARE
Natural Internal 1.2
₹42,300.00INR / Case of 500
DWK Life Sciences (Wheaton) CryoELITE® Cryogenic Vials, Sterile, Polypropylene, Internal Thread, Round Bottom, Natural Cap, 2 mL; 500/Cs WW-03756-27 Mfr # W985911
COMPARE
Natural Internal 2
₹45,800.00INR / Case of 500
DWK Life Sciences (Wheaton) CryoELITE® Cryogenic Vials, Sterile, Polypropylene, Internal Thread, Flat Bottom, White Cap, 2 mL; 500/Cs WW-03756-29 Mfr # W985916
COMPARE
White Internal 2
₹50,200.00INR / Case of 500
DWK Life Sciences (Wheaton) CryoELITE® Cryogenic Vials, Sterile, Polypropylene, External Thread, Flat Bottom, Red Cap, 2 mL; 500/Cs WW-03756-31 Mfr # W985864
COMPARE
Red External 2
₹50,100.00INR / Case of 500
DWK Life Sciences (Wheaton) CryoELITE® Cryogenic Vials, Sterile, Polypropylene, External Thread, Flat Bottom, Pink Cap, 2 mL; 500/Cs WW-03756-33 Mfr # W985865
COMPARE
Pink External 2
₹50,100.00INR / Case of 500
DWK Life Sciences (Wheaton) CryoELITE® Cryogenic Vials, Sterile, Polypropylene, Internal Thread, Round Bottom, Natural Cap, 4 mL; 500/Cs WW-03756-37 Mfr # W985913
COMPARE
Natural Internal 4
₹63,400.00INR / Case of 500
Showing 1- 10 of 10