WHEATON® Liquid Scintillation Vials, Glass
Clear vials allow for each sample viewing
  • Converted from Type I borosilicate glass tubing
These 20-mL scintillation vials are made of Type I borosilicate glass. Vials feature background counts are consistent and low and ultraviolet transmission is high, making them ideal for solvent reduction. Multiple vial and cap configuration options are available for a wide range of applications. Vials are supplied with caps attached.
49,400.00 - ₹53,600.00INR / Case of 500
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Cap Material Septum Type Thread
Availability Pricing
DWK Life Sciences (Wheaton) Liquid Scintillation Vials, Glass, Urea Cap with Metal Foil/Pulp Liner, 22-400 Thread, 20 mL; 500/Cs WW-08918-01 Mfr # 986542
COMPARE
Urea Metal Foil / Pulp 22-400
₹49,400.00INR / Case of 500
DWK Life Sciences (Wheaton) Liquid Scintillation Vials, Glass, Urea Cap with Polyethylene Cone, 22-400 Thread, 20 mL; 500/Cs WW-08918-02 Mfr # 986546
COMPARE
Urea Polyethylene Cone 22-400
₹53,600.00INR / Case of 500
DWK Life Sciences (Wheaton) Liquid Scintillation Vials, Glass, Urea Cap with Polyethylene Disc, 22-400 Thread, 20 mL; 500/Cs WW-08918-03 Mfr # 986548
COMPARE
Urea Polyethylene Disc 22-400
₹51,800.00INR / Case of 500
DWK Life Sciences (Wheaton) Liquid Scintillation Vials, Glass, Urea Cap with Polyethylene Disc, 24-400 Thread, 20 mL; 500/Cs WW-08918-05 Mfr # 986568
COMPARE
Urea Polyethylene Disc 24-400
₹51,800.00INR / Case of 500
DWK Life Sciences (Wheaton) Liquid Scintillation Vials, Glass, Polypropylene Cap with Metal Foil/Pulp Liner, 22-400 Thread, 20 mL; 500/Cs WW-08918-06 Mfr # 986541
COMPARE
Polypropylene Metal Foil / Pulp 22-400