Weksler Low-Cost Dual Scale Utility Gauges
Quality gauge is designed for general pressure applications
  • Use with automated assembly, agricultural, and process equipment
For your day to day pressure measurement needs, these quality dry gauges are an economical solution for a variety of applications. Gauges feature a white background with kPa in blue on outer scale and psi in black on the inner ring. Case and ring is black finish steel with plastic lens. Case is black steel with a plastic window. Tube is bronze, socket is brass.
1,200.00 - ₹1,400.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Weksler Dual Scale Gauge, 1.5" Dial, 30"/0" VAC / 100 to 0 kPa, 1/8" NPT Bottom Connection WW-68345-00 Mfr # UA15H8L
COMPARE
Dual Scale Gauge, 1.5\" Dial, 30\"/0\" VAC / 100 to 0 kPa, 1/8\" NPT Bottom Connection
₹1,300.00INR / Each
Weksler Dual Scale Gauge, 1.5" Dial, 0 to 100 psi / 0 to 700 kPa, 1/8" NPT Bottom Connection WW-68345-08 Mfr # UA15C8L
COMPARE
Dual Scale Gauge, 1.5\" Dial, 0 to 100 psi / 0 to 700 kPa, 1/8\" NPT Bottom Connection
₹1,200.00INR / Each
Weksler Dual Scale Gauge, 1.5" Dial, 0 to 160 psi / 0 to 1100 kPa, 1/8" NPT Bottom Connection WW-68345-10 Mfr # UA15D8L
COMPARE
Dual Scale Gauge, 1.5\" Dial, 0 to 160 psi / 0 to 1100 kPa, 1/8\" NPT Bottom Connection
₹1,200.00INR / Each
Weksler Dual Scale Gauge, 1.5" Dial, 30"/0" VAC / 100 to 0 kPa, 1/8" NPT Back Connection WW-68345-14 Mfr # UA15H8C
COMPARE
Dual Scale Gauge, 1.5\" Dial, 30\"/0\" VAC / 100 to 0 kPa, 1/8\" NPT Back Connection
₹1,400.00INR / Each
Weksler Dual Scale Gauge, 1.5" Dial, 0 to 15 psi / 0 to 100 kPa, 1/8" NPT Back Connection WW-68345-16 Mfr # UA15N8C
COMPARE
Dual Scale Gauge, 1.5\" Dial, 0 to 15 psi / 0 to 100 kPa, 1/8\" NPT Back Connection
₹1,300.00INR / Each
Weksler Dual Scale Gauge, 1.5" Dial, 0 to 60 psi / 0 to 400 kPa, 1/8" NPT Back Connection WW-68345-20 Mfr # UA15B8C
COMPARE
Dual Scale Gauge, 1.5\" Dial, 0 to 60 psi / 0 to 400 kPa, 1/8\" NPT Back Connection
₹1,300.00INR / Each
Weksler Dual Scale Gauge, 1.5" Dial, 0 to 100 psi / 0 to 700 kPa, 1/8" NPT Back Connection WW-68345-22 Mfr # UA15C8C
COMPARE
Dual Scale Gauge, 1.5\" Dial, 0 to 100 psi / 0 to 700 kPa, 1/8\" NPT Back Connection
₹1,300.00INR / Each
Showing 1- 7 of 7