ITS WaterWorks Water Quality Test Strips
Obtain test results within 90 seconds
  • Test strips feature bright easy-to-read colors
  • No hazardous reagents
These economical water quality test strips are formulated to test tap water, lake or stream water, rain run-off water, or treated water.
2,700.00 - ₹8,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Measurement Points Test Tests Per Kit
Availability Pricing
ITS 480008 Water Quality Test Strips, total hardness, 0 to 425 ppm, 50/bottle WW-99531-03 Mfr # 480008
COMPARE
0 to 425 ppm Hardness, total 50
₹2,700.00INR / Each
ITS 480115 Water Quality Test Strips, Multi-Test Strip. WW-99531-15 Mfr # 480115
COMPARE
Free chlorine, Total chlorine, pH, Total alkalinity and, Total hardness pH, free and total chlorine, hardness, alkalinity 1
₹3,900.00INR / Pkg of 30
ITS 481026 Water Quality Test Strips, Free Chlorine, 0 to 5.0 ppm, 50/Bottle WW-99531-20 Mfr # 481026
COMPARE
0, 0.05, 0.1, 0.2, 0.4, 0.5, 0.8, 1.0, 2.0, 5.0 ppm Chlorine, free 50
₹5,200.00INR / Each
ITS 480010 Water Quality Test Strips, total chlorine, 0 to 10.0 ppm, 50/bottle WW-99531-22 Mfr # 480010
COMPARE
0, 0.05, 0.075, 0.1, 0.125, 0.15, 0.175, 0.2, 0.3, 0.5, 0.8, 4.0, 10.0 ppm Chlorine, total 50
₹8,000.00INR / Each
ITS 480005 Water Quality Test Strips, PH/Total Alkalinity (Ta), 6-9 PH/0-360 Ta, 50/Bottle WW-99531-28 Mfr # 480005
COMPARE
6 to 9 pH; 0, 80, 120, 180, 240, 360 TA pH and total alkalinity 50
₹3,200.00INR / Each
ITS 480014 Water Quality Test Strips, peroxide, 0 to 100 ppm, 50/bottle WW-99531-30 Mfr # 480014
COMPARE
0, 0.5, 2.0, 5.0, 10, 25, 50, 100 ppm Peroxide 50
₹4,200.00INR / Each